Learn More
Tisab Solution, Pure, Fisher Chemical™
Product Code. 10162140 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10162140 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10162140 Supplier Fisher Chemical Supplier No. F/1325/21
Request Bulk or Custom Format
Tisab Solution, Pure, Fisher Chemical, Quantity: 5L, Packaging: Plastic container, Colorless, pH: 5 to 7, MDL Number: 148650, Physical Form: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
pH 5 to 7
Physical Form Liquid
Quantity 5 L
Formulation Sodium Chloride, Water, 1,2-Cyclohexanediamine, Sodium Acetate
MDL Number 148650
Packaging Plastic container
Solubility Information Miscible