missing translation for 'onlineSavingsMsg'
Learn More

Tirabrutinib hydrochloride, MedChemExpress

Product Code. p-200100195
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499357

Brand: Medchem Express HY15771A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tirabrutinib (ONO-4059) hydrochloride is an orally active Bruton’s Tyrosine Kinase (BTK) inhibitor (can cross the blood-brain barrier (BBB)), with an IC50 of 6.8 nM. Tirabrutinib hydrochloride irreversibly and covalently binds to BTK and inhibits aberrant B cell receptor signaling. Tirabrutinib hydrochloride can be used in studies of autoimmune diseases and hematological malignancies.

Tirabrutinib (ONO-4059) hydrochloride is an orally active Bruton’s Tyrosine Kinase (BTK) inhibitor (can cross the blood-brain barrier (BBB)), with an IC50 of 6.8 nM. Tirabrutinib hydrochloride irreversibly and covalently binds to BTK and inhibits aberrant B cell receptor signaling. Tirabrutinib hydrochloride can be used in studies of autoimmune diseases and hematological malignancies.

Specifications

Chemical Name or Material Tirabrutinib hydrochloride
CAS 1439901-97-9
Color White
Purity Grade Notes Research
Molecular Formula C25H23ClN6O3
Quantity 5 mg
Synonym ONO-4059 hydrochlorideGS-4059 hydrochloride
Solubility Information DMSO : 100 mg/mL (203.69 mM; Need ultrasonic)
SMILES O=C1N(C2=CC=C(OC3=CC=CC=C3)C=C2)C4=C(N)N=CN=C4N1[C@H]5CN(C(C#CC)=O)CC5.[H]Cl
Molecular Weight (g/mol) 490.94
Formula Weight 490.94
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.