missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tirabrutinib hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Tirabrutinib (ONO-4059) hydrochloride is an orally active Bruton’s Tyrosine Kinase (BTK) inhibitor (can cross the blood-brain barrier (BBB)), with an IC50 of 6.8 nM. Tirabrutinib hydrochloride irreversibly and covalently binds to BTK and inhibits aberrant B cell receptor signaling. Tirabrutinib hydrochloride can be used in studies of autoimmune diseases and hematological malignancies.
Specifications
Specifications
| Chemical Name or Material | Tirabrutinib hydrochloride |
| CAS | 1439901-97-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H23ClN6O3 |
| Quantity | 5 mg |
| Synonym | ONO-4059 hydrochlorideGS-4059 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (203.69 mM; Need ultrasonic) |
| SMILES | O=C1N(C2=CC=C(OC3=CC=CC=C3)C=C2)C4=C(N)N=CN=C4N1[C@H]5CN(C(C#CC)=O)CC5.[H]Cl |
| Molecular Weight (g/mol) | 490.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction