missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tipranavir, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Tipranavir (PNU-140690) inhibits the enzymatic activity and dimerization of HIV-1 protease, exerts potent activity against multi-protease inhibitor (PI)-resistant HIV-1 isolates with IC50s of 66-410 nM. Tipranavir inhibits SARS-CoV-2 3CLpro activity.
Specifications
Specifications
| Chemical Name or Material | Tipranavir |
| CAS | 174484-41-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H33F3N2O5S |
| Quantity | 1 mg |
| Synonym | PNU-140690 |
| Solubility Information | DMSO : 200 mg/mL (331.86 mM; Need ultrasonic) ∣Ethanol : ≥ 50 mg/mL (82.97 mM) |
| SMILES | O=C1C([C@H](CC)C2=CC=CC(NS(C3=CC=C(C(F)(F)F)C=N3)(=O)=O)=C2)=C(O)C[C@](CCC4=CC=CC=C4)(CCC)O1 |
| Molecular Weight (g/mol) | 602.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction