Learn More
Lovibond™ Tintometer™ DPD No. 3 High Calcium Tablets
Product Code. 15301975
Click to view available options
Quantity:
100 Tablets
250 Tablets
500 Tablets
Unit Size:
Pack of 100
Pack of 250
Pack of 500
Product Code. 15301975
Brand: Lovibond™ 515731BT
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Especificaciones
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type DPD No. 3 High Calcium Tablets
compliance-icons
Nombre del producto
  • X250 DPD NO.3 HIGH CALCIUM TABLETS .
Palabra de advertencia
  • Danger
Clasificación
  • Specific target organ toxicity Category 1
Indicaciones de peligro
  • H372-Causes damage to organs.
Consejos de prudencia
  • P264-Wash hands thoroughly after handling.
  • P314-Get medical advice/attention if you feel unwell.
Otros peligros
  • MIXTURE LIST-Contains : potassium iodide
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado