Learn More
Lovibond™ Tintometer™ Chloride Tablets
Product Code. 15321995
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
Product Code. 15321995
Brand: Lovibond™ 502461
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Chloride Tablets
  • Tablet Count Method
  • Bottle of 250
Specifications
For Use With (Application) Photometry Reagent
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Test Method Tablet Count Method
Type Chloride Tablets
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • X250 CHLORIDE TABLETS
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, potassium chromate, silver nitrate, potassium dichromate