missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator Test Kit, Ks 4.3
Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
- Grade: Ks4.3
Specifications
Specifications
| Accuracy | ±5% FS |
| Description | Grade: Ks4.3 |
| For Use With (Application) | Water Quality Testing |
| For Use With (Equipment) | Checkit Comprator Tests |
| Includes | Carry case with comparator, vials, discs, reagents and method instructions for all tests |
| Range | 0.5 to 5mmol/L |
| Type | Checkit Comprator Test Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?