missing translation for 'onlineSavingsMsg'
Learn More

Melford Timentin

Product Code. p-200087208
Click to view available options
Quantity:
2 g
5 g
10 g
25 g
Unit Size:
10g
25g
2g
5g
This item is not returnable. View return policy

Product Code. 30018853

Brand: Melford T360005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor).

Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor). The combination of ticarcillin and clavulanate enhances the spectrum of antibacterial activity by protecting ticarcillin from degradation by certain bacterial enzymes.

Ticarcillin inhibits bacterial cell wall synthesis, disrupting the integrity of the bacterial cell wall and leading to bacterial cell lysis. Clavulanate potassium works by inhibiting beta-lactamases, enzymes produced by bacteria that can inactivate beta-lactam antibiotics, such as ticarcillin. This combination is effective against a broad spectrum of bacteria, including those producing beta-lactamases.

Specifications

Chemical Name or Material Timentin
CAS 4697-14-7, 61177-45-5
Quantity 5 g
Molecular Formula C15H14N2Na2O6S2/C8H8KNO5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.