missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Rat anti-Mouse, Clone-826118, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB7267-SP
This item is not returnable.
View return policy
Description
TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
| TIGIT | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 201633 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Blockade of Receptor-ligand Interaction | |
| 826118 | |
| Blockade of Receptor-ligand Interaction 40-240 ng/mL | |
| P86176 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIGIT Met1-Thr143 Accession # P86176 | |
| 25 μg | |
| Primary | |
| Detects mouse TIGIT in direct ELISAs. | |
| Mouse | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction