missing translation for 'onlineSavingsMsg'
Learn More

Tianeptine Sodium Salt, TRC

Product Code. 17950935
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950935 100 mg 100mg
17900895 500 mg 500mg
17900385 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17950935 Supplier Toronto Research Chemicals Supplier No. TRCT436805100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30123-17-2
Molecular Formula C21 H24 Cl N2 O4 S . Na
Molecular Weight (g/mol) 458.93
Synonym Tianeptine sodium, Sodium 7-[[(11RS)-3-chloro-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepin-11-yl]amino]heptanoate S,S-dioxide, 7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt (1:1), Heptanoic acid, 7-[(3-chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]-, monosodium salt (9CI), Heptanoic acid, 7-[(3-chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]-, S,S-dioxide, monosodium salt (8CI), Dibenzo[c,f][1,2]thiazepine, heptanoic acid deriv., Sodium 7-[(3-chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoate S,S-dioxide, Stablon, Tianeptine sodium, Tianeptine sodium salt
IUPAC Name sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)amino]heptanoate
SMILES [Na+].CN1c2ccccc2C(NCCCCCCC(=O)[O-])c3ccc(Cl)cc3S1(=O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H25ClN2O4S.Na/c1-24-18-9-6-5-8-16(18)21(23-13-7-3-2-4-10-20(25)26)17-12-11-15(22)14-19(17)29(24,27)28;/h5-6,8-9,11-12,14,21,23H,2-4,7,10,13H2,1H3,(H,25,26);/q;+1/p-1
Formula Weight 458.1043
Chemical Name or Material Sodium Tianeptine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.