missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tianeptine Sodium, Mikromol™
Description
Tianeptine Sodium
Specifications
Specifications
| Chemical Name or Material | Sodium Tianeptine |
| CAS | 30123-17-2 |
| Analyte or Component Names | Sodium Tianeptine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 Cl N2 O4 S . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tianeptine sodium, Sodium 7-[[(11RS)-3-chloro-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepin-11-yl]amino]heptanoate S,S-dioxide, 7-[(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic acid sodium salt (1:1), Heptanoic acid, 7-[(3-chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)amino]-, monosodium salt (9CI), Heptanoic acid, 7-[(3-chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]-, S,S-dioxide, monosodium salt (8CI), Dibenzo[c,f][1,2]thiazepine, heptanoic acid deriv., Sodium 7-[(3-chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoate S,S-dioxide, Stablon, Tianeptine sodium, Tianeptine sodium salt |
| InChI Formula | InChI=1S/C21H25ClN2O4S.Na/c1-24-18-9-6-5-8-16(18)21(23-13-7-3-2-4-10-20(25)26)17-12-11-15(22)14-19(17)29(24,27)28;/h5-6,8-9,11-12,14,21,23H,2-4,7,10,13H2,1H3,(H,25,26);/q;+1/p-1 |
| SMILES | [Na+].CN1c2ccccc2C(NCCCCCCC(=O)[O-])c3ccc(Cl)cc3S1(=O)=O |
| IUPAC Name | sodium;7-[(3-chloro-6-methyl-5,5-dioxo-11H-benzo[c][1,2]benzothiazepin-11-yl)amino]heptanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?