missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine 4’-O-Sulfate Disodium Salt, Thyroxine 4’-O-Sulfate Disodium Salt, TRC

Product Code. 30011169
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011169 2.5 mg 2.5mg
30009065 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011169 Supplier Toronto Research Chemicals Supplier No. TRCT4256382.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H9I4NNa2O7S
Molecular Weight (g/mol) 900.9
Synonym O-[3,5-Diiodo-4-(sulfooxy)phenyl]-3,5-diiodo-L-tyrosine, Thyroxine Hydrogen Sulfate, Thyroxine Sulfate Disodium Salt;
SMILES N[C@H](C(O[Na])=O)CC(C=C1I)=CC(I)=C1OC2=CC(I)=C(OS(=O)(O[Na])=O)C(I)=C2

Specifications

Analyte or Component Names Thyroxine 4'-O-Sulfate Disodium Salt
Shipping Condition Room Temperature
Industry Type Forensics
Quantity 2.5 mg
Percent Purity >95%
Chemical Name or Material Thyroxine 4'-O-Sulfate Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.