Learn More
Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.
154.02 EUR - 1085.00 EUR
Specifications
| Antigen | Thrombospondin-1 (TSP-1) Ab-2 |
|---|---|
| Clone | D4.6 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Form | Concentrated |
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|
12691377
|
Epredia
MS-419-B0 |
100 μL | Biotin | 200μg/mL | |||||
|
12611387
|
Epredia
MS-419-B1 |
500 μL | Biotin | 200μg/mL | |||||
|
12601387
|
Epredia
MS-419-P0 |
100 μL | Unconjugated | 200μg/mL | |||||
|
12631387
|
Epredia
MS-419-P1 |
500 μL | Unconjugated | 200μg/mL | |||||
|
12621387
|
Epredia
MS-419-P |
1 mL | Unconjugated | 200μg/mL | |||||
|
12641387
|
Epredia
MS-419-P1ABX |
100 μL | Unconjugated | 1mg/mL | |||||
|
12661387
|
Epredia
MS-419-PABX |
200 μL | Unconjugated | 1mg/mL | |||||
Description
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Host Species: Mouse
Clone: D4.6
Isotype: IgG1
Species Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Dog and Sheep. Others not known.
Epitope: Calcium-binding domain of TSP (C-terminal 50kDa piece of the 120kDa fragment)
Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunoprecipitation (Native verified)
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
| Thrombospondin-1 (TSP-1) Ab-2 | |
| Immunoprecipitation, Western Blot | |
| Concentrated | |
| RUO | |
| Bovine, Canine, Equine, Human, Mouse, Pig, Rat, Sheep | |
| IgG1 |
| D4.6 | |
| Monoclonal | |
| Mouse | |
| Angiogenesis | |
| Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.