missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Threo-Metaraminol Fumarate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Threo-Metaraminol Fumarate Salt |
| Analyte or Component Names | Threo-Metaraminol Fumarate Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9H13NO2 • x(C4H4O4) |
| Synonym | Threo-α-(1-aminoethyl)-m-hydroxy-benzyl Alcohol Fumarate Salt, (R*,R*)-α-(1-Aminoethyl)-3-hydroxy-benzenemethanol Fumarate Salt |
| InChI Formula | InChI=1S/C9H13NO2.C4H4O4/c1-6(10)9(12)7-3-2-4-8(11)5-7;5-3(6)1-2-4(7)8/h2-6,9,11-12H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t6-,9+;/m0./s1 |
| Solution Type | Neat |
| SMILES | O=C(/C=C/C(O)=O)O.OC1=CC=CC([C@@H]([C@H](C)N)O)=C1 |
| IUPAC Name | 3-((1S,2S)-2-amino-1-hydroxypropyl)phenol fumarate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?