missing translation for 'onlineSavingsMsg'
Learn More

Threo-Metaraminol Fumarate Salt, TRC

Product Code. 30449666
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449666 5 mg 5mg
30450080 50 mg 50mg
30453749 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449666 Supplier Toronto Research Chemicals Supplier No. TRCT4045025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Threo-Metaraminol Fumarate Salt
Analyte or Component Names Threo-Metaraminol Fumarate Salt
Shipping Condition Room Temperature
Molecular Formula C9H13NO2 • x(C4H4O4)
Synonym Threo-α-(1-aminoethyl)-m-hydroxy-benzyl Alcohol Fumarate Salt, (R*,R*)-α-(1-Aminoethyl)-3-hydroxy-benzenemethanol Fumarate Salt
InChI Formula InChI=1S/C9H13NO2.C4H4O4/c1-6(10)9(12)7-3-2-4-8(11)5-7;5-3(6)1-2-4(7)8/h2-6,9,11-12H,10H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t6-,9+;/m0./s1
Solution Type Neat
SMILES O=C(/C=C/C(O)=O)O.OC1=CC=CC([C@@H]([C@H](C)N)O)=C1
IUPAC Name 3-((1S,2S)-2-amino-1-hydroxypropyl)phenol fumarate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 283.28
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.