Learn More
Codan™ Three-part Syringes
Use Codan™ Three-part Syringes to accommodate a variety of medical needs including: catheterization, hypodermic ear treatments, and more. The syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spill
Brand: Codan™ 62.2611
Additional Details : Weight : 0.27000kg
Description
- Sterile
- Graduations
- Disposable
- Latex-free
Specifications
Autoclavable | |
Dual scale; Silicone seal | |
Yes | |
2 mL | |
Yes |
Polypropylene | |
Hypodermic | |
0.1 mL | |
Centric Luer Cone | |
Sterile |