Learn More
Cole-Parmer™ Threaded Nipple NPT(M) Fitting
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-31
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11878053
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Threaded Nipple Fitting
Autoclavable
3/4 in.
Autoclave or Ethylene Oxide
Translucent
NPT(M) Connection
PFA
3/4 in.
3/4 in. NPT(M)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions