Learn More
Cole-Parmer™ Threaded Nipple NPT(M) Fitting
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-26
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11858043
  • 39.20 EUR / Each
Estimated Shipment: 17-06-2024
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Threaded Nipple Fitting
Autoclavable
1 13/32 in.
3/8 in.
3/8 in. NPT(M)
Translucent
NPT(M) Connection
PFA
3/8 in.
Autoclave or Ethylene Oxide
13/32 in.