Learn More
Cole-Parmer™ Threaded Nipple NPT(M) Fitting
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-24
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 11770389
  • 42.95 EUR / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Threaded Nipple Fitting
Autoclavable
1 13/32 in.
1/4 in.
1/4 in. NPT(M)
Translucent
NPT(M) Connection
PFA
1/4 in.
Autoclave or Ethylene Oxide
5/8 in.