Learn More
Buerkle™ Thread Adapters PP
Shop All Bürkle Products
Click to view available options
Color:
Black
Black, Orange
Colorless
Gray
Violet
Description
- Constructed of polypropylene
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Polypropylene |
| Thread Size | External: S61x6 (DIN61), Internal: G2 in. |
| Color | Gray |
| Product Type | Thread Adapter |