missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-carbonitrile, 98%

Product Code. 11463914
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11463914

Brand: Thermo Scientific Alfa Aesar A11930.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1003-31-2 | C5H3NS | 109.146 g/mol

Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-31-2
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00005416
InChI Key CUPOOAWTRIURFT-UHFFFAOYSA-N
Synonym 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile
PubChem CID 66087
IUPAC Name thiophene-2-carbonitrile
SMILES C1=CSC(=C1)C#N

Specifications

Density 1.171
Boiling Point 191°C to 193°C
Flash Point 81°C (178°F)
Odor Unpleasant
Refractive Index 1.5645
Quantity 100 g
UN Number UN3276
Beilstein 107035
Solubility Information Slightly miscible with water.
Formula Weight 109.15
Percent Purity 98%
Chemical Name or Material Thiophene-2-carbonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.