Description
- Available in mixed or natural colors
- Available in individual tubes or strips
- Available with a variety of caps - domed, flat, strip
Specifications
Specifications
| Color | Assorted |
| Volume (Metric) | 0.2 mL |
| Closure Material | Polypropylene |
| For Use With (Equipment) | PCR |
| Closure Type | Flat Cap |
| Format | Individual |
| Product Type | PCR Tube |