Learn More
Thiamet G, MedChemExpress
Product Code. 16427977
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16427977
Brand: Medchem Express HY12588
MedChemExpress Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.
Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.
Specifications
Chemical Name or Material Thiamet G
Physical Form Powder
CAS 1009816-48-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H16N2O4S
For Use With (Application) Cancer-Kinase/protease
Solubility Information H2O : ≥ 50 mg/mL (201.37 mM) ∣DMSO : ≥ 45 mg/mL (181.23 mM)
SMILES O[C@H]1[C@H](O)[C@@]2([H])N=C(NCC)S[C@@]2([H])O[C@@H]1CO
Molecular Weight (g/mol) 248.3
Formula Weight 248.3
Percent Purity 98.32%
Grade Research