Learn More
Velp Scientifica™ Thermostatic Bath
Product Code. 11706394 Shop All Velp Products
Click to view available options
Length (Metric):
410 mm
Unit Size:
Each
Product Code. 11706394
Brand: Velp Scientifica™ A00001055
Only null left
Add to basket
Add to basket
Velp Scientifica™ Thermostatic Bath for use with MULTISTIRRER and MULTISTIRRER DIGITAL.
  • For use with MULTISTIRRER and MULTISTIRRER DIGITAL
Specifications
For Use With (Equipment) MULTISTIRRER and MULTISTIRRER DIGITAL
Height (Metric) 180 mm
Length (Metric) 410 mm
Type Thermostatic Bath
Width (Metric) 280 mm