missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit

Product Code. 15823374
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15823374

Brand: Applied Biosystems™ 792601KT

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Thermo Sequenase with thermostable inorganic Pyrophosphatase, Reaction Buffer (concentrate), Control DNA pUC19, Control Primer, TAMRA -40 Universal Forward Primer, ddG Termination Mix (7-deaza-dGTP), ddA Termination Mix (7-deaza-dGTP), ddT Termination Mix (7-deaza-dGTP), ddC Termination Mix (7-deaza-dGTP), Formamide Loading Dye, Protocol Book

The Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit is designed to be used with fluorescent dye-labeled primers and high-resolution fluorescence scanners.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit Components:
Thermo Sequenase with thermostable inorganic Pyrophosphatase
Reaction Buffer (concentrate)
Control DNA pUC19
Control Primer, TAMRA -40 Universal Forward Primer
ddG Termination Mix (7-deaza-dGTP)
ddA Termination Mix (7-deaza-dGTP)
ddT Termination Mix (7-deaza-dGTP)
ddC Termination Mix (7-deaza-dGTP)
Formamide Loading Dye
Protocol Book

The kit and all the enclosed reagents should be stored frozen at -20°C (NOT in a frost-free freezer). Keep all reagents on ice when removed from storage for use.
Format Kit
For Use With (Application) Sequencing
Primer Universal Forward Primer
Product Type Thermo Sequenase Dye Primer Manual Cycle Sequencing Kit
Quantity 1 Kit

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.