Learn More
Epredia™ Lab Vision™ UltraVision™ Quanto Mouse on Mouse HRP Detection System
HRP polymer designed to detect mouse antibodies on mouse tissue, maximizing signal while minimizing background.
Brand:  Epredia™ TL-015-QHDM
Additional Details : Weight : 0.14969kg
 View more versions of this product
Product Code. 15270715
  • 3120.00 EUR / 15mL
Estimated Shipment: 06-06-2024
Description
Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Epredia UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely.
UltraVision Quanto Makes Optimal Results Routine by providing:
  • Strong Staining
  • Reduced Mouse on Mouse Background
  • Simple Biotin Free Polymer Procedure
  • For use with a DAB or other HRP Chromogen; For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL)
Specifications
Specifications
15 mL