Learn More
Thermo Scientific™ Heracell™ Support Frame (780 mm high)
Product Code. 10719992 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Heracell 150i CO2 incubator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10719992 Heracell 150i CO2 incubator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10719992 Supplier Thermo Scientific™ Supplier No. 50051436
Only null left
Add to basket
Add to basket
For Use With: Heracell 150i CO2 Incubator
  • Provide protection against floor contamination
  • Choice of three heights: 185mm (7.3 in.), 200mm (7.9 in.), or 780mm (30.7 in.)
  • Support frames can also be caster-mounted for easy maneuverability
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heracell 150i CO2 incubator
Lighting LED
Type Support Frame
Height (English) 30.7 in.
Height (Metric) 78 cm