Description
For use with Thermo Scientific Sorvall Stratos Centrifuges.
Specifications
Specifications
| Type | Microtiter Plate Rotor |
| Capacity | 2 x 1 mL |
| Max. RCF | 1,370 x g |
| Max. Speed | 3,000 rpm |
| Radius (Metric) | 1.36 cm |
| For Use With (Equipment) | Thermo Scientific™ Sorvall™ Stratus Centrifuge |
| Certifications/Compliance | Biocontainment approved |