Learn More
Nunc™ Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
Click to view available options
Holds:
2 mL Cryogenic Tubes
5 mL Cryogenic Tubes
Unit Size:
Pack of 10
Description
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes.
- Non-sterile
- Case of 10
Specifications
Specifications
| For Use With (Equipment) | For 5mL Cryobank, non-sterile |
| Holds | 5 mL Cryogenic Tubes |
| Quantity | 10/pk., 10/cs. |
| Type | Cryogenic Tube Storage Rack (w/Latch) |