Learn More
Thermo Scientific™ Microplate Readers Optical Filters
Brand:  Thermo Scientific™ 1425700
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11819540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • All filters have a 3nm bandpass centered at the specified wavelength
  • Compatible with all three Multiskan models
Specifications
Specifications
570 nm