Learn More
Thermo Scientific™ Microplate Readers Optical Filters
197.00 EUR - 724.00 EUR
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11562389
|
Thermo Scientific™
1425200 |
520 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11809540
|
Thermo Scientific™
1425500 |
550 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11819540
|
Thermo Scientific™
1425700 |
570 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11849540
|
Thermo Scientific™
1426500 |
650 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10389773
|
Thermo Scientific™
1424051 |
405 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11749428
|
Thermo Scientific™
1425600 |
560 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- All filters have a 3nm bandpass centered at the specified wavelength
- Compatible with all three Multiskan models