Learn More
Thermo Scientific™ Microplate Readers Optical Filters
Brand: Thermo Scientific™ 1425200
Additional Details : Weight : 0.10000kg
Description
- All filters have a 3nm bandpass centered at the specified wavelength
- Compatible with all three Multiskan models
Specifications
520 nm |