missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Microplate Readers Optical Filters
Brand: Thermo Scientific™ 1424051
Additional Details : Weight : 0.80000kg
Description
- All filters have a 3nm bandpass centered at the specified wavelength
- Compatible with all three Multiskan models
Specifications
405 nm |