Learn More
Thermo Scientific™ Labtips Pipette Tips
Description
- Double injection molded for maximum sample delivery and best fit for a variety of pipetters
- Available in 200 and 1000μL including bulk and racked formats
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Sterility | Non-sterile |
| Color | Blue |
| Autoclavable | Autoclavable |
| Format | Rack |
| Tip Style | Universal Fit |
| For Use With (Equipment) | LabTip Pipette |
| Product Line | LabTip |
| Material | Polypropylene |