Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Product Code. 12823412 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Capacity:
108 x 15 mL Blood Collection Tube
132 x 7 mL Blood Collection Tubes
18 x 50 mL Tubes
30 x 50 mL Tubes
48 x 15 mL Conical Blood Collection Tube
5 mL Blood Collection Tubes
6 x 250 mL Bottles
6 x 500 mL Bottles
6 x 550 mL Blood Bags
missing translation for 'unitSize'
Each
Product Code. 12823412
missing translation for 'mfr': Thermo Scientific™ 00445
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
TRUSTED_SUSTAINABILITY
Specifications
Capacity 18 x 50 mL Tubes
For Use With (Equipment) H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor