Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Rectangular Longpath Cell Holder
Shop All Thermo Scientific ProductsDescription
Rectangular Longpath Cell Holder. Easy-to-install “snap-in” cell holder accommodates one rectangular longpath cell up to 50mm long and 12mm wide.
Specifications
Specifications
| Type | Cell Holder |
| For Use With (Equipment) | Genesys™ 20 spectrophotometer |
| Holds | Cell up to 50m L and 12mm W |
| Pathlength | 50 mm |
| Shape | Rectangular |