Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel HLT 5000uL Pipette Tips
Description
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
Specifications
Specifications
| Volume (Metric) | 5000 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Format | Bulk |
| Tip Style | Blunt |
| Product Line | ART |