Learn More
WTW™ Thermal Matrix Printer
Click to view available options
Type:
Thermal Matrix Printer
Unit Size:
Each
Description
- As transfer rates for most of the devices are fixed, the printer might need to be adjusted to the matching transfer rate
- WTW™ devices with serial interfaces can be connected directly to a computer and therefore be used to print to a printer
- 40 characters per line
- For paper width 112mm, thermos printer paper, normal quality, readable for approx.. 5 years; high quality readable for at least 10 years
Specifications
Specifications
| Description | 40 characters per line |
| Type | Thermal Matrix Printer |
| Electrical Requirements | 230 VAC, 50 Hz and battery operation |
| Width (Metric) | 112 mm paper |