Learn More
WTW™ Thermal Matrix Printer
Product Code. 10163871
Click to view available options
Type:
Thermal Matrix Printer
Unit Size:
Each
Product Code. 10163871
Brand: WTW™ 250045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Equipped with a serial RS 232 interface and can be connected directly to the PC or the appropriate devices.
  • As transfer rates for most of the devices are fixed, the printer might need to be adjusted to the matching transfer rate
  • WTW™ devices with serial interfaces can be connected directly to a computer and therefore be used to print to a printer
  • 40 characters per line
  • For paper width 112mm, thermos printer paper, normal quality, readable for approx.. 5 years; high quality readable for at least 10 years
TRUSTED_SUSTAINABILITY
Specifications
Description 40 characters per line
Type Thermal Matrix Printer
Electrical Requirements 230 VAC, 50 Hz and battery operation
Width (Metric) 112 mm paper