Learn More
LTE Scientific™ Thermal Data Printer
Thermal Data Printer for LTE Touchlave models.
1200.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12735385
|
LTE Scientific™
TC/TDP/01 |
All LTE Touchclave K & F Model Autoclaves |
1200.00 EUR
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
12735405
|
LTE Scientific™
TC/TDP/02 |
All LTE Touchclave R Model Autoclaves |
1200.00 EUR
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
Description
- Thermal data printer
- Prints all cycle data as cycle progresses
- Printout includes any faults/alerts
- Provides printed validation of load success
Specifications
Thermal Data Printer | |
Non-sterile |