Learn More
LTE Scientific™ Thermal Data Printer
Click to view available options
For Use With (Equipment):
LTE Touchclave K and F Autoclaves
LTE Touchclave R Autoclaves
Unit Size:
Each
Description
- Thermal data printer
- Prints all cycle data as cycle progresses
- Printout includes any faults/alerts
- Provides printed validation of load success
Specifications
Specifications
| For Use With (Equipment) | LTE Touchclave K and F Autoclaves |
| Model | TC/TDP/01 |
| Product Type | Thermal Data Printer |
| Sterility | Non-sterile |