missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thalidomide-O-amido-C4-NH2 TFA, MedChemExpress
Description
Thalidomide-O-amido-C4-NH2 TFA (Cereblon Ligand-Linker Conjugates 6 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Specifications
Specifications
| Chemical Name or Material | Thalidomide-O-amido-C4-NH2 TFA |
| CAS | 1799711-25-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H23F3N4O8 |
| Quantity | 100 mg |
| Synonym | Cereblon Ligand-Linker Conjugates 6 TFAE3 ligase Ligand-Linker Conjugates 19 TFA |
| Solubility Information | DMSO : 42.86 mg/mL (82.99 mM; Need ultrasonic) |
| SMILES | NCCCCNC(COC1=C(C(N2C(CCC3=O)C(N3)=O)=O)C(C2=O)=CC=C1)=O.FC(F)(F)C(O)=O |
| Molecular Weight (g/mol) | 516.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?