missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thalidomide-O-amido-C4-NH2 TFA, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Description
Thalidomide-O-amido-C4-NH2 TFA (Cereblon Ligand-Linker Conjugates 6 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Specifications
Specifications
| Chemical Name or Material | Thalidomide-O-amido-C4-NH2 TFA |
| CAS | 1799711-25-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H23F3N4O8 |
| Quantity | 1 g |
| Synonym | Cereblon Ligand-Linker Conjugates 6 TFAE3 ligase Ligand-Linker Conjugates 19 TFA |
| Solubility Information | DMSO : 42.86 mg/mL (82.99 mM; Need ultrasonic) |
| SMILES | NCCCCNC(COC1=C(C(N2C(CCC3=O)C(N3)=O)=O)C(C2=O)=CC=C1)=O.FC(F)(F)C(O)=O |
| Molecular Weight (g/mol) | 516.42 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction