Learn More
Thalidomide-O-amido-C4-N3, MedChemExpress
Product Code. 16494916
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Product Code. 16494916
Brand: Medchem Express HY103615
MedChemExpress Thalidomide-O-amido-C4-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Thalidomide-O-amido-C4-N3 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Specifications
Chemical Name or Material Thalidomide-O-amido-C4-N3
CAS 2098488-36-7
Color White
Purity Grade Notes Research
Molecular Formula C19H20N6O6
Quantity 100 mg
Synonym Cereblon Ligand-Linker Conjugates 4 E3 ligase Ligand-Linker Conjugates 18
Solubility Information DMSO : ≥ 41 mg/mL (95.70 mM)
SMILES O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(NCCCCN=[N+]=[N-])=O
Molecular Weight (g/mol) 428.4
Formula Weight 428.4
Percent Purity 98.63%
Grade Research
Physical Form Solid