missing translation for 'onlineSavingsMsg'
Learn More

TH5487, MedChemExpress

Product Code. 16485238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485238 10 mg 10mg
16477957 5 mg 5mg
16415916 100 mg 100mg
16401457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485238 Supplier Medchem Express Supplier No. HY125276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TH5487 is a potent 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor with an IC50 of 342 nM. TH5487 stops OGG1 from recognizing its DNA substrate, inhibits DNA repair and modifies OGG1 chromatin dynamics, which results in the inhibition of proinflammatory pathway genes.

TH5487 is a potent 8-oxoguanine DNA glycosylase 1 (OGG1) inhibitor with an IC50 of 342 nM. TH5487 stops OGG1 from recognizing its DNA substrate, inhibits DNA repair and modifies OGG1 chromatin dynamics, which results in the inhibition of proinflammatory pathway genes.

Specifications

Chemical Name or Material TH5487
CAS 2304947-71-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H18BrIN4O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (46.20 mM; Need ultrasonic)
SMILES O=C(N1CCC(N(C2=CC=CC(Br)=C2N3)C3=O)CC1)NC4=CC=C(I)C=C4
Molecular Weight (g/mol) 541.18
Formula Weight 541.18
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.