Learn More
Thermo Scientific™ TH-641 Swinging Bucket Rotor Package for WX+ Ultraspeed Centrifuges
Product Code. 12161690 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
6 x 13.2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12161690 6 x 13.2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12161690 Supplier Thermo Scientific™ Supplier No. 54295
Process samples effectively with the TH-641 Swinging Bucket Rotor.
Process samples effectively with the Thermo Scientific™ TH-641 Swinging Bucket Rotor Package.
Ordering Information:
Order ultracrimp/clearcrimp sealing tool (03920), crimp gauge replacement (03919), extra plugs and caps (03045), tube removal tool (12826), tube racks (two) (03726) separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Sorvall WX Ultra Series Floor Model Centrifuge
Recommended for:
Rate zonal isolation of RNA and subcellular particles
TRUSTED_SUSTAINABILITY
Specifications
Description TH-641 Swinging Bucket Rotor Package
Material Titanium
Type Swinging Bucket Rotor
Capacity 6 x 13.2 mL
Max. RCF 287,660 x g
Max. Speed 41,000 rpm
Radius (Metric) 153.2 mm
Radius (Metric) Min. 71.9 mm
K Factor 114
Max. Tube Length 89 mm
Max. Tube Diameter 14 mm
Net Weight 12.3 lb. (5.6 kg)
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle 90°
Avg. RCF 211,333 x g
Min. RCF 135,005 x g