missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120515
This item is not returnable.
View return policy
Description
Positive test controls include: HepG2, MCF7, K-562, U-251MG, Mouse liver, Mouse testis, Rat uterus. Immunogen sequence: SSVESVMGIK NFMPALEETP FHSCTAGPNP TLGRPLSPKP SSPGSVLARP SVICHTTVTA LKDVPFSLCQ SVGVGQNTDI QQIAAKNFTD T.
TGIF is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain.
Specifications
| TGIF | |
| Polyclonal | |
| Unconjugated | |
| TGIF1 | |
| 5'-TG-3'-interacting factor 1; AA959811; AI462167; Homeobox protein TGIF1; HPE4; TALE family homeobox; TALE homeobox TG-interacting factor; Tfig; TG interacting factor 1; TGFB induced factor homeobox 1; TGFB-induced factor homeobox 1; TGIF; Tgif1; transforming growth factor-beta-induced factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21815, 316742, 7050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P70284, Q15583 | |
| TGIF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-340 of human TGIF1 (NP_733796.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction