missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGIF Polyclonal Antibody

Description
Predicted to react with chicken based on sequence homology.
TGIF is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain.
Specifications
Specifications
| Antigen | TGIF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TGIF1 |
| Gene Accession No. | P70284, Q15583 |
| Gene Alias | 5'-TG-3'-interacting factor 1; AA959811; AI462167; Homeobox protein TGIF1; HPE4; TALE family homeobox; TALE homeobox TG-interacting factor; Tfig; TG interacting factor 1; TGFB induced factor homeobox 1; TGFB-induced factor homeobox 1; TGIF; Tgif1; transforming growth factor-beta-induced factor |
| Gene Symbols | TGIF1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?