missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TGF-beta 1 Magnetic Luminex™ Performance Assay
Beskrivelse
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Tekniske data
Tekniske data
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Multi-species |
| Binding Target | TGF-beta 1 |
| Bead Type | Magnetic |
| Cross Reactivity | Human, Mouse, Rat, Porcine |
| Gene ID (Entrez) | 7040 |
| Synonym | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Kit Contents |
|
| Content And Storage | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?