missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18835-1-AP-150UL
This item is not returnable.
View return policy
Description
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5′-GCCNNNGGC-3′ and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.Specifications
| TFAP2D | |
| Polyclonal | |
| Unconjugated | |
| TFAP2D | |
| AP2 delta, TFAP2BL1, TFAP2D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 226896, 301284, 83741 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z6R9, Q91ZK0 | |
| Tfap2d | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction