missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl Diphosphine Disulfide, MP Biomedicals

Product Code. 11499364 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 11499364

Brand: MP Biomedicals 0522249983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3676-97-9
Molecular Formula C4H12P2S2
Quantity 250 mg
Synonym tetramethyldiphosphine disulfide, tetramethylbiphosphine disulfide, diphosphine, tetramethyl-, 1,2-disulfide, 1,1,2,2-tetramethyldiphosphane 1,2-disulfide, dimethylphosphinothioyl-dimethyl-thioxo-$l^ 5-phosphane, tetramethyl-1??,2??-diphosphane-1,2-dithione, diphosphine, tetramethyl-, disulfide, teramethyldiphosphine disulphide, tetramethyldiphosphoryl disulfide, 1,2-bisthiotetramethyldiphosphorane
InChI Key LGZQQMXQPAHRIH-UHFFFAOYSA-N
SMILES CP(=S)(C)P(=S)(C)C
IUPAC Name dimethylphosphinothioyl-dimethyl-sulfanylidene-$l^{5}-phosphane
Molecular Weight (g/mol) 186.208
PubChem CID 96660
Formula Weight 185.21
Chemical Name or Material Tetramethyl Diphosphine Disulfide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.